siRNA Details
Gene Symbol: Ldb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107601.1 | NP_001101071.1 | 2 | 579 | ||
XM_006231493.1 | XP_006231555.1 | 2 | 411 | |||
XM_006231494.1 | XP_006231556.1 | 2 | 411 | |||
GenBank mRNA | BC166757.1 | - | 2 | 216 |
