siRNA Details
Gene Symbol: Gbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191634.1 | NP_001178563.1 | 34, 35 | 4844 | ||
XM_006231496.2 | XP_006231558.1 | 35, 36 | 5020 | |||
XM_006231497.2 | XP_006231559.1 | 35, 36 | 5017 | |||
XM_006231498.2 | XP_006231560.1 | 35, 36 | 5008 | |||
XM_006231500.2 | XP_006231562.1 | 34, 35 | 4921 | |||
XM_006231501.2 | XP_006231563.1 | 34, 35 | 4909 | |||
XM_006231502.2 | XP_006231564.1 | 34, 35 | 4906 |
