siRNA Details
Gene Symbol: Gbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191634.1 | NP_001178563.1 | 25, 26 | 3428 | ||
XM_006231496.2 | XP_006231558.1 | 26, 27 | 3604 | |||
XM_006231497.2 | XP_006231559.1 | 26, 27 | 3601 | |||
XM_006231498.2 | XP_006231560.1 | 26, 27 | 3604 | |||
XM_006231500.2 | XP_006231562.1 | 25, 26 | 3505 | |||
XM_006231501.2 | XP_006231563.1 | 25, 26 | 3505 | |||
XM_006231502.2 | XP_006231564.1 | 25, 26 | 3502 |
