siRNA Details
Gene Symbol: Sorcs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191563.1 | NP_001178492.1 | 14 | 1859 | ||
XM_006231519.2 | XP_006231581.1 | 14 | 2082 | |||
XM_006231520.2 | XP_006231582.1 | 14 | 2082 | |||
XM_006231521.2 | XP_006231583.1 | 14 | 2082 | |||
XM_006231522.2 | XP_006231584.1 | 14 | 2082 | |||
XM_006231523.2 | XP_006231585.1 | 14 | 2082 | |||
XM_006231524.2 | XP_006231586.1 | 14 | 2082 | |||
XM_006231525.1 | XP_006231587.1 | 14 | 2082 | |||
XM_008760441.1 | XP_008758663.1 | 14 | 1545 | |||
XM_008760442.1 | XP_008758664.1 | 14 | 1704 |
