siRNA Details
Gene Symbol: Pofut2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107621.2 | NP_001101091.2 | 2 | 430 | ||
XM_006256299.2 | XP_006256361.1 | 2 | 1025 | |||
GenBank mRNA | CV103570.1 | - | 1 | 162 |
