siRNA Details
Gene Symbol: Bcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_228091.9 | XP_228091.6 | 14 | 2967 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_228091.9 | XP_228091.6 | 14 | 2967 |