siRNA Details
Gene Symbol: Rev3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083966.1 | NP_001077435.1 | 13 | 5793 | ||
XM_006256524.1 | XP_006256586.1 | 13 | 5798 | |||
XM_006256525.2 | XP_006256587.1 | 14 | 5556 | |||
XM_006256527.2 | XP_006256589.1 | 13 | 5798 | |||
XM_008772987.1 | XP_008771209.1 | 13 | 5859 | |||
XM_008772988.1 | XP_008771210.1 | 13 | 5798 |
