siRNA Details
Gene Symbol: Qrsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014034.1 | NP_001014056.1 | 2 | 208 | ||
XM_006256588.2 | XP_006256650.1 | 2 | 247 | |||
GenBank mRNA | BC089213.1 | - | 2 | 208 | 7302358 |
