siRNA Details
Gene Symbol: Pde8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199268.1 | NP_954889.1 | 4, 5 | 387 | ||
XM_006231811.2 | XP_006231873.1 | 5, 6 | 1001 | |||
XM_008760669.1 | XP_008758891.1 | 5, 6 | 998 | |||
XM_008760670.1 | XP_008758892.1 | 5, 6 | 1001 | |||
GenBank mRNA | AB092697.1 | - | 4, 5 | 387 |
