siRNA Details
Gene Symbol: Ptcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107648.1 | NP_001101118.1 | 5 | 494 | ||
XM_006231847.2 | XP_006231909.1 | 5 | 526 | |||
XM_008760675.1 | XP_008758897.1 | 4 | 1716 |
