siRNA Details
Gene Symbol: Zc2hc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107661.1 | NP_001101131.1 | 5 | 477 | ||
XM_006232144.2 | XP_006232206.1 | 5 | 496 | |||
GenBank mRNA | BC168976.1 | - | 5 | 439 | 9033370 |
