siRNA Details
Gene Symbol: Phc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107662.1 | NP_001101132.1 | 11, 12 | 2491 | ||
XM_006232226.2 | XP_006232288.1 | 14, 15 | 2713 | |||
XM_006232227.2 | XP_006232289.1 | 13, 14 | 2665 | |||
XM_006232229.2 | XP_006232291.1 | 13, 14 | 2614 | |||
XM_006232230.2 | XP_006232292.1 | 12, 13 | 2566 |
