siRNA Details
Gene Symbol: Hps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107664.1 | NP_001101134.1 | 2 | 401 | ||
XM_008760868.1 | XP_008759090.1 | 2 | 956 | |||
XM_008760869.1 | XP_008759091.1 | 2 | 956 | |||
XM_008760870.1 | XP_008759092.1 | 2 | 955 | |||
XM_008760871.1 | XP_008759093.1 | 2 | 955 |
