siRNA Details
Gene Symbol: Nim1k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231982.2 | XP_006232044.1 | 2 | 2676 | ||
XM_008760801.1 | XP_008759023.1 | 2 | 1190 | |||
XM_227081.6 | XP_227081.3 | 2 | 1806 |
