siRNA Details
Gene Symbol: Nmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107682.2 | NP_001101152.1 | 7, 8 | 709 | ||
XM_008761090.1 | XP_008759312.1 | 7, 8 | 704 | |||
GenBank mRNA | BC161998.1 | - | 7, 8 | 709 | 9035992 | |
FQ215207.1 | - | 5, 6 | 446 |
