siRNA Details
Gene Symbol: Slitrk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107683.1 | NP_001101153.1 | 2 | 460 | ||
XM_006232490.2 | XP_006232552.1 | 2 | 730 | |||
XM_008761091.1 | XP_008759313.1 | 2 | 202 |
