siRNA Details
Gene Symbol: Rapgef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107684.1 | NP_001101154.1 | 5 | 774 | ||
XM_006232511.2 | XP_006232573.2 | 9 | 2662 | |||
XM_006232512.2 | XP_006232574.1 | 7 | 1728 | |||
XM_006232513.2 | XP_006232575.1 | 7 | 1539 | |||
XM_008761092.1 | XP_008759314.1 | 13 | 2032 | |||
XM_008761093.1 | XP_008759315.1 | 12 | 2015 | |||
XM_008761094.1 | XP_008759316.1 | 13 | 2032 | |||
XM_008761095.1 | XP_008759317.1 | 13 | 1706 | |||
XM_008761096.1 | XP_008759318.1 | 11 | 2321 | |||
XM_008761097.1 | XP_008759319.1 | 10 | 2213 | |||
XM_008761098.1 | XP_008759320.1 | 10 | 3505 |
