siRNA Details
Gene Symbol: Rapgef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107684.1 | NP_001101154.1 | 19 | 3442 | ||
XM_006232511.2 | XP_006232573.2 | 24 | 5354 | |||
XM_006232512.2 | XP_006232574.1 | 22 | 4420 | |||
XM_006232513.2 | XP_006232575.1 | 22 | 4231 | |||
XM_008761092.1 | XP_008759314.1 | 28 | 4724 | |||
XM_008761093.1 | XP_008759315.1 | 27 | 4707 | |||
XM_008761094.1 | XP_008759316.1 | 27 | 4700 | |||
XM_008761095.1 | XP_008759317.1 | 28 | 4398 | |||
XM_008761096.1 | XP_008759318.1 | 26 | 5013 | |||
XM_008761097.1 | XP_008759319.1 | 25 | 4905 | |||
XM_008761098.1 | XP_008759320.1 | 25 | 6197 |
