siRNA Details
Gene Symbol: Tlr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198769.2 | NP_942064.1 | 2 | 2130 | ||
XM_008761102.1 | XP_008759324.1 | 3 | 2495 | |||
GenBank mRNA | AY151255.1 | - | 2 | 2130 | ||
AY151256.1 | - | 2 | 2130 |
