siRNA Details
Gene Symbol: Gatad2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024888.1 | NP_001020059.1 | 6 | 982 | ||
XM_008761144.1 | XP_008759366.1 | 6 | 913 | |||
XM_008761145.1 | XP_008759367.1 | 6 | 913 | |||
GenBank mRNA | BC097428.1 | - | 6 | 982 | 7455294 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_139304, AK141586, AK173113, BC130262, AK041594, and others.
Gene Ontology Categories:
