siRNA Details
Gene Symbol: Gatad2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024888.1 | NP_001020059.1 | 11 | 1921 | ||
XM_008761144.1 | XP_008759366.1 | 11 | 1852 | |||
XM_008761145.1 | XP_008759367.1 | 11 | 1849 | |||
GenBank mRNA | BC097428.1 | - | 11 | 1921 | 7455294 |
