siRNA Details
Gene Symbol: Sema4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012078.2 | NP_001012078.2 | 13 | 1715 | ||
XM_006232639.2 | XP_006232701.1 | 13 | 1595 | |||
XM_006232640.2 | XP_006232702.1 | 13 | 1566 | |||
XM_006232641.2 | XP_006232703.1 | 13 | 1795 | |||
XM_008761150.1 | XP_008759372.1 | 13 | 2289 | |||
GenBank mRNA | BC081943.1 | - | 13 | 1659 | 7121310 |
