siRNA Details
Gene Symbol: Trim46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107691.1 | NP_001101161.1 | 6 | 1009 | ||
XM_008761170.1 | XP_008759392.1 | 6 | 1126 | |||
XM_008761171.1 | XP_008759393.1 | 6 | 973 | |||
XM_008761172.1 | XP_008759394.1 | 6 | 972 | |||
XM_008761173.1 | XP_008759395.1 | 6 | 973 |
Gene Symbol: LOC103690106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773704.1 | XP_008771926.1 | 1 | 288 |
