siRNA Details
Gene Symbol: Tdrkh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014038.2 | NP_001014060.2 | 5 | 621 | ||
XM_006232870.2 | XP_006232932.1 | 5 | 624 | |||
XM_006232871.2 | XP_006232933.1 | 5 | 2580 | |||
GenBank mRNA | BC059161.1 | - | 5 | 577 | 6887770 |
