siRNA Details
Gene Symbol: Tdrkh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014038.2 | NP_001014060.2 | 3 | 306 | ||
XM_006232870.2 | XP_006232932.1 | 3 | 309 | |||
XM_006232871.2 | XP_006232933.1 | 3 | 2265 | |||
GenBank mRNA | BC059161.1 | - | 3 | 262 | 6887770 |
