siRNA Details
Gene Symbol: LOC100363361 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729081.4 | XP_002729127.1 | 9 | 3834 | ||
XM_006232933.2 | XP_006232995.1 | 9 | 3910 | |||
XM_006232934.2 | XP_006232996.1 | 10 | 3977 | |||
XM_006232935.2 | XP_006232997.1 | 9 | 3696 | |||
XM_008761341.1 | XP_008759563.1 | 9 | 6155 |
