siRNA Details
Gene Symbol: LOC100363361 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729081.4 | XP_002729127.1 | 3 | 2468 | ||
XM_006232933.2 | XP_006232995.1 | 3 | 2544 | |||
XM_006232934.2 | XP_006232996.1 | 4 | 2611 | |||
XM_006232935.2 | XP_006232997.1 | 3 | 2330 | |||
XM_008761341.1 | XP_008759563.1 | 3 | 4789 |
