siRNA Details
Gene Symbol: Tnfaip8l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014039.1 | NP_001014061.1 | 3 | 447 | ||
XM_008761303.1 | XP_008759525.1 | 2 | 1200 | |||
GenBank mRNA | BC079019.1 | - | 3 | 447 | 7113794 | |
FQ219780.1 | - | 2 | 158 |
