siRNA Details
Gene Symbol: Prune | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007697.1 | NP_001007698.1 | 8 | 1581 | ||
XM_006232885.2 | XP_006232947.1 | 7 | 1434 | |||
GenBank mRNA | BC079054.1 | - | 8 | 1581 | 7115417 |
