siRNA Details
Gene Symbol: Setdb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271175.1 | NP_001258104.1 | 16 | 2864 | ||
XM_006232913.2 | XP_006232975.1 | 17 | 3254 | |||
XM_006232914.2 | XP_006232976.1 | 16 | 3035 | |||
XM_006232916.2 | XP_006232978.1 | 12 | 2231 | |||
XM_006232917.2 | XP_006232979.1 | 12 | 2734 |
