siRNA Details
Gene Symbol: Adamtsl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034012.1 | NP_001029184.1 | 1 | 160 | ||
GenBank mRNA | BC099119.1 | - | 1 | 160 | 7462597 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adamtsl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034012.1 | NP_001029184.1 | 1 | 160 | ||
GenBank mRNA | BC099119.1 | - | 1 | 160 | 7462597 |