siRNA Details
Gene Symbol: Tars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014040.1 | NP_001014062.1 | 2 | 124 | ||
XM_008761308.1 | XP_008759530.1 | 3 | 2643 | |||
GenBank mRNA | BC079154.1 | - | 2 | 124 | 7128703 |
