siRNA Details
Gene Symbol: Mov10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107711.1 | NP_001101181.1 | 8 | 1489 | ||
XM_006233085.1 | XP_006233147.1 | 8 | 1377 | |||
XM_008761395.1 | XP_008759617.1 | 8 | 2237 |
