siRNA Details
Gene Symbol: Fndc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107717.1 | NP_001101187.1 | 5 | 927 | ||
XM_006233159.2 | XP_006233221.1 | 6 | 935 | |||
XM_006233160.2 | XP_006233222.1 | 6 | 928 |
