siRNA Details
Gene Symbol: Extl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100704.1 | NP_001094174.1 | 3 | 501 | ||
XM_006233215.2 | XP_006233277.1 | 3 | 1046 | |||
XM_006233216.2 | XP_006233278.1 | 3 | 787 | |||
XM_006233217.2 | XP_006233279.1 | 2 | 578 | |||
GenBank mRNA | BC162014.1 | - | 3 | 501 | 9091843 |
