siRNA Details
Gene Symbol: Fnbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039609.1 | NP_001034698.1 | 1218 | |||
GenBank mRNA | AB250295.1 | - | 1218 | |||
AB250296.1 | - | 1044 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fnbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039609.1 | NP_001034698.1 | 1218 | |||
GenBank mRNA | AB250295.1 | - | 1218 | |||
AB250296.1 | - | 1044 |