siRNA Details
Gene Symbol: Zzz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134549.2 | NP_001128021.2 | 6 | 2092 | ||
XM_006233489.2 | XP_006233551.1 | 8 | 2376 | |||
XM_006233490.2 | XP_006233552.1 | 9 | 2966 | |||
XM_006233491.2 | XP_006233553.1 | 8 | 2373 | |||
XM_008761551.1 | XP_008759773.1 | 9 | 2405 | |||
GenBank mRNA | FM103952.1 | - | 4 | 281 | ||
FN806367.1 | - | 3 | 255 |

More Information
Other Species Targeted

Gene Ontology Categories:
