siRNA Details
Gene Symbol: Zzz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134549.2 | NP_001128021.2 | 10 | 2568 | ||
XM_006233489.2 | XP_006233551.1 | 12 | 2852 | |||
XM_006233490.2 | XP_006233552.1 | 13 | 3442 | |||
XM_006233491.2 | XP_006233553.1 | 12 | 2849 | |||
XM_008761551.1 | XP_008759773.1 | 13 | 2881 |
