siRNA Details
Gene Symbol: Usp33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191094.1 | NP_001178023.1 | 5, 6 | 454 | ||
XM_006233492.2 | XP_006233554.1 | 6, 7 | 619 | |||
XM_006233493.2 | XP_006233555.1 | 6, 7 | 619 |
