siRNA Details
Gene Symbol: Usp33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191094.1 | NP_001178023.1 | 16 | 1804 | ||
XM_006233492.2 | XP_006233554.1 | 17 | 1993 | |||
XM_006233493.2 | XP_006233555.1 | 17 | 1969 |
