siRNA Details
Gene Symbol: Slc35c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107748.1 | NP_001101218.1 | 1 | 723 | ||
XM_006234568.2 | XP_006234630.1 | 2 | 932 | |||
XM_008762026.1 | XP_008760248.1 | 2 | 948 |
