siRNA Details
Gene Symbol: Ext2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107751.1 | NP_001101221.1 | 10 | 1737 | ||
XM_006234576.2 | XP_006234638.2 | 10 | 1766 | |||
GenBank mRNA | BC166748.1 | - | 10 | 1724 | 5599208 | |
FQ218956.1 | - | 1 | 16 |
