siRNA Details
Gene Symbol: Casc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170594.1 | NP_001164065.1 | 7 | 793 | ||
XM_008762095.1 | XP_008760317.1 | 10 | 979 | |||
XM_008762096.1 | XP_008760318.1 | 10 | 1209 | |||
XM_008762097.1 | XP_008760319.1 | 10 | 1022 | |||
XM_008762098.1 | XP_008760320.1 | 8 | 892 | |||
XM_008762099.1 | XP_008760321.1 | 10 | 966 |
Gene Symbol: LOC100911204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234821.2 | XP_006234883.1 | 979 | |||
XM_006234822.2 | XP_006234884.1 | 1209 | ||||
XM_006234823.2 | XP_006234885.1 | 967 | ||||
XM_008762127.1 | XP_008760349.1 | 1027 | ||||
XM_008762128.1 | XP_008760350.1 | 892 |
