siRNA Details
Gene Symbol: Rpap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033999.2 | NP_001029171.1 | 23 | 3946 | ||
XM_006234771.2 | XP_006234833.1 | 23 | 4003 | |||
XM_006234772.2 | XP_006234834.1 | 22 | 3892 | |||
XM_006234773.2 | XP_006234835.1 | 22 | 3871 |
