siRNA Details
Gene Symbol: Ttbk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107766.1 | NP_001101236.1 | 11 | 1470 | ||
XM_006234833.2 | XP_006234895.1 | 13 | 1903 | |||
XM_006234834.2 | XP_006234896.1 | 13 | 1546 | |||
XM_006234835.2 | XP_006234897.1 | 12 | 1829 | |||
XM_006234836.2 | XP_006234898.1 | 11 | 1681 | |||
XM_006234837.2 | XP_006234899.1 | 13 | 1549 |
