siRNA Details
Gene Symbol: Pax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107787.1 | NP_001101257.1 | 2 | 1064 | ||
XM_006235152.2 | XP_006235214.2 | 3 | 2370 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107787.1 | NP_001101257.1 | 2 | 1064 | ||
XM_006235152.2 | XP_006235214.2 | 3 | 2370 |