siRNA Details
Gene Symbol: Itch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005887.1 | NP_001005887.1 | 7 | 814 | ||
XM_008762335.1 | XP_008760557.1 | 9 | 1042 | |||
XM_008762336.1 | XP_008760558.1 | 9 | 967 | |||
XM_008762337.1 | XP_008760559.1 | 6 | 719 | |||
GenBank mRNA | AY600518.1 | - | 7 | 814 |
