siRNA Details
Gene Symbol: Myh7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107794.2 | NP_001101264.2 | 40 | 5829 | ||
XM_008762338.1 | XP_008760560.1 | 41 | 6690 | |||
XM_008762339.1 | XP_008760561.1 | 41 | 6096 | |||
XM_008762340.1 | XP_008760562.1 | 41 | 5833 | |||
XM_008762341.1 | XP_008760563.1 | 41 | 6690 | |||
GenBank mRNA | EU241478.1 | - | 40 | 5829 |
