siRNA Details
Gene Symbol: Chd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107797.1 | NP_001101267.1 | 31 | 5554 | ||
XM_006235482.2 | XP_006235544.1 | 30 | 5834 | |||
XM_006235483.2 | XP_006235545.1 | 30 | 6250 | |||
XM_006235484.2 | XP_006235546.1 | 29 | 5768 | |||
XM_008762345.1 | XP_008760567.1 | 31 | 5613 | |||
XM_008762346.1 | XP_008760568.1 | 30 | 5558 |
