siRNA Details
Gene Symbol: L3mbtl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_230849.7 | XP_230849.6 | 17, 18 | 3049 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: L3mbtl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_230849.7 | XP_230849.6 | 17, 18 | 3049 |